Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magee2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106941.1 | NP_001100411.1 | 1 | 1142 | ||
XM_006257144.2 | XP_006257206.1 | 3 | 2495 | |||
XM_008773341.1 | XP_008771563.1 | 2 | 2682 | |||
XM_008773342.1 | XP_008771564.1 | 3 | 2555 |