Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102550809 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256902.2 | XP_006256964.1 | 5 | 1542 | ||
XM_008773212.1 | XP_008771434.1 | 4 | 3000 |