Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tns4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024881.1 | NP_001020052.1 | 4 | 1174 | ||
GenBank mRNA | BC097378.1 | - | 4 | 1174 | 7445075 |
Gene Symbol: NEWGENE_1310402 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768229.1 | XP_008766451.1 | 4 | 878 |