Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif18b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039019.1 | NP_001034108.1 | 6 | 909 | ||
XM_006247505.2 | XP_006247567.1 | 6 | 917 | |||
XM_008768283.1 | XP_008766505.1 | 6 | 918 | |||
GenBank mRNA | BC099126.1 | - | 6 | 909 | 7455959 |