Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agfg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107131.1 | NP_001100601.1 | 4 | 663 | ||
XM_006249140.2 | XP_006249202.1 | 5 | 843 | |||
XM_006249141.2 | XP_006249203.1 | 5 | 842 | |||
XM_006249142.2 | XP_006249204.1 | 5 | 844 |