Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107138.1 | NP_001100608.1 | 14 | 1777 | ||
XM_006249300.2 | XP_006249362.1 | 14 | 2354 | |||
XM_006249301.2 | XP_006249363.1 | 14 | 2351 | |||
XM_008769224.1 | XP_008767446.1 | 13 | 2249 | |||
XM_008769225.1 | XP_008767447.1 | 13 | 2246 | |||
XM_008769226.1 | XP_008767448.1 | 11 | 1259 | |||
GenBank mRNA | FQ230302.1 | - | 5 | 466 | ||
FQ232681.1 | - | 5 | 466 | |||
FQ233202.1 | - | 4 | 365 |