Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alkbh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126273.1 | NP_001119745.1 | 1 | 664 | ||
XM_006249498.2 | XP_006249560.1 | 2 | 403 | |||
GenBank mRNA | BC166547.1 | - | 1 | 664 | 7304677 |