Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Depdc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107229.1 | NP_001100699.1 | 12 | 864 | ||
XM_006251277.2 | XP_006251339.1 | 13 | 1036 | |||
XM_006251279.2 | XP_006251341.1 | 13 | 1036 | |||
XM_006251281.2 | XP_006251343.1 | 13 | 1036 | |||
XM_006251283.2 | XP_006251345.1 | 13 | 1036 | |||
XM_008770289.1 | XP_008768511.1 | 12 | 874 | |||
XM_008770290.1 | XP_008768512.1 | 13 | 1008 | |||
XM_008770291.1 | XP_008768513.1 | 13 | 992 | |||
XM_008770292.1 | XP_008768514.1 | 12 | 862 | |||
XM_008770293.1 | XP_008768515.1 | 13 | 1036 |