Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751423.3 | XP_003751471.1 | 21 | 4143 | ||
XM_006251620.2 | XP_006251682.1 | 20 | 3999 | |||
XM_008770473.1 | XP_008768695.1 | 21 | 3875 |