Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728176.4 | XP_002728222.1 | 2 | 791 | ||
XM_006251719.2 | XP_006251781.1 | 2 | 2295 |