Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107254.1 | NP_001100724.1 | 6, 7 | 1562 | ||
XM_008770515.1 | XP_008768737.1 | 6, 7 | 2439 | |||
XM_008770516.1 | XP_008768738.1 | 5, 6 | 2174 | |||
GenBank mRNA | BC168894.1 | - | 6, 7 | 1583 | 7453870 |