Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107254.1 | NP_001100724.1 | 5 | 1266 | ||
XM_008770515.1 | XP_008768737.1 | 5 | 2104 | |||
XM_008770516.1 | XP_008768738.1 | 4 | 1839 | |||
GenBank mRNA | BC168894.1 | - | 5 | 1248 | 7453870 |