Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271342.1 | NP_001258271.1 | 3 | 1409 | ||
XM_006254228.2 | XP_006254290.1 | 3 | 1409 |