Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spire1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107381.1 | NP_001100851.1 | 5 | 831 | ||
XM_006254860.2 | XP_006254922.1 | 6 | 1061 | |||
XM_006254862.2 | XP_006254924.1 | 6 | 1061 | |||
XM_006254863.2 | XP_006254925.1 | 6 | 1060 | |||
XM_006254864.2 | XP_006254926.1 | 6 | 1060 |