Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dock1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143858.1 | NP_001137330.1 | 5, 6 | 436 | ||
XM_008759987.1 | XP_008758209.1 | 5, 6 | 429 |