Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728854.4 | XP_002728900.1 | 3 | 750 | ||
XM_006230930.2 | XP_006230992.1 | 3 | 751 | |||
XM_008760191.1 | XP_008758413.1 | 1 | 77 | |||
XM_008760192.1 | XP_008758414.1 | 1 | 74 |