Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100909704 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760552.1 | XP_008758774.1 | 1 | 164 |
Gene Symbol: Ccdc147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749139.3 | XP_003749187.1 | 16 | 2524 |
Gene Symbol: Cfap58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231594.2 | XP_006231656.1 | 16 | 2524 |