Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749139.3 | XP_003749187.1 | 5 | 866 |
Gene Symbol: Cfap58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231594.2 | XP_006231656.1 | 5 | 866 |