Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014033.1 | NP_001014055.1 | 3 | 371 | ||
XM_006256415.2 | XP_006256477.1 | 4 | 596 | |||
XM_006256417.2 | XP_006256479.1 | 3 | 589 | |||
XM_006256418.2 | XP_006256480.1 | 4 | 595 | |||
XM_008772886.1 | XP_008771108.1 | 3 | 874 | |||
GenBank mRNA | BC087157.1 | - | 3 | 371 | 7133379 |