Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232927.2 | XP_006232989.1 | 21 | 3692 | ||
XM_006232928.2 | XP_006232990.1 | 21 | 3775 | |||
XM_006232929.2 | XP_006232991.1 | 21 | 3693 | |||
XM_006232930.2 | XP_006232992.1 | 21 | 3801 |