Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100702.1 | NP_001094172.1 | 4, 5 | 540 | ||
XM_006233157.2 | XP_006233219.1 | 4, 5 | 673 |