Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spg11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191571.1 | NP_001178500.1 | 7, 8 | 1577 | ||
XM_006234855.2 | XP_006234917.1 | 7, 8 | 1932 |