Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107830.1 | NP_001101300.1 | 14 | 2532 | ||
XM_008761668.1 | XP_008759890.1 | 14 | 2615 | |||
XM_008761669.1 | XP_008759891.1 | 14 | 2615 |