Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238446.2 | XP_006238508.1 | 13 | 1989 | ||
XM_006238447.2 | XP_006238509.1 | 13 | 1893 | |||
XM_006238448.2 | XP_006238510.1 | 13 | 3053 | |||
XM_006238449.2 | XP_006238511.1 | 14 | 3116 |