Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf113a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014791.1 | NP_001014791.1 | 340 | |||
GenBank mRNA | AY363108.1 | - | 344 | |||
BC087595.1 | - | 340 | 7310345 |
Gene Symbol: Rnf113a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773490.1 | XP_008771712.1 | 1 | 362 |