Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107970.1 | NP_001101440.1 | 1 | 148 | ||
XM_006238759.2 | XP_006238821.1 | 1 | 192 | |||
XM_006238760.2 | XP_006238822.1 | 1 | 190 | |||
XM_006238761.2 | XP_006238823.1 | 1 | 193 |