Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rap1gap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100713.1 | NP_001094183.1 | 21 | 1898 | ||
XM_006239189.1 | XP_006239251.1 | 20 | 1975 | |||
XM_006239193.1 | XP_006239255.1 | 19 | 1897 | |||
GenBank mRNA | BC089979.1 | - | 5 | 440 |