Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35e2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107998.1 | NP_001101468.1 | 3 | 1242 | ||
XM_006239572.2 | XP_006239634.1 | 3 | 804 | |||
XM_006239573.2 | XP_006239635.1 | 3 | 921 | |||
XM_008764361.1 | XP_008762583.1 | 3 | 921 |