Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankmy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108019.1 | NP_001101489.1 | 1 | 243 | ||
XM_006240043.2 | XP_006240105.2 | 2 | 2945 |