Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108038.1 | NP_001101508.1 | 21 | 3861 | ||
XM_008764780.1 | XP_008763002.1 | 21 | 3903 | |||
XM_008764781.1 | XP_008763003.1 | 21 | 3897 |