Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creb3l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012115.1 | NP_001012115.1 | 2, 3 | 227 | ||
GenBank mRNA | BC089877.1 | - | 2, 3 | 227 | 7325489 |