Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubash3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191792.1 | NP_001178721.1 | 14 | 1932 | ||
XM_006242848.2 | XP_006242910.1 | 14 | 2251 | |||
XM_006242850.2 | XP_006242912.1 | 14 | 2506 | |||
XM_008766123.1 | XP_008764345.1 | 14 | 2260 | |||
GenBank mRNA | FQ221553.1 | - | 14 | 2183 | ||
FQ223998.1 | - | 14 | 2183 |