Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pih1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191793.1 | NP_001178722.1 | 5 | 848 | ||
XM_006243028.2 | XP_006243090.1 | 4 | 614 | |||
XM_006243029.2 | XP_006243091.1 | 3 | 521 | |||
XM_006243030.2 | XP_006243092.1 | 3 | 520 |