Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127490.1 | NP_001120962.1 | 9 | 2040 | ||
XM_006243326.2 | XP_006243388.1 | 10 | 1953 | |||
XM_008766291.1 | XP_008764513.1 | 11 | 1917 | |||
XM_008766292.1 | XP_008764514.1 | 11 | 1601 |