Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sphkap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127492.1 | NP_001120964.1 | 8 | 1500 | ||
XM_006245230.2 | XP_006245292.1 | 8 | 1672 | |||
XM_006245231.2 | XP_006245293.1 | 8 | 2073 | |||
XM_006245233.2 | XP_006245295.1 | 7 | 1592 | |||
XM_006245234.2 | XP_006245296.1 | 8 | 1671 | |||
XM_008767251.1 | XP_008765473.1 | 7 | 1994 |