Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map7d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271327.1 | NP_001258256.1 | 10 | 1663 | ||
XM_008773649.1 | XP_008771871.1 | 10 | 1835 | |||
XM_008773650.1 | XP_008771872.1 | 10 | 1747 |