Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750899.3 | XP_003750947.1 | 2 | 341 | ||
XM_006247119.2 | XP_006247181.1 | 2 | 567 | |||
XM_006247120.2 | XP_006247182.1 | 2 | 343 |