Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Helq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014134.2 | NP_001014156.2 | 6, 7 | 1623 | ||
XM_006250666.2 | XP_006250728.1 | 6, 7 | 1620 | |||
XM_008770040.1 | XP_008768262.1 | 4, 5 | 449 |