Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252959.2 | XP_006253021.1 | 6 | 1111 | ||
XM_006252960.2 | XP_006253022.1 | 6 | 1110 | |||
XM_006252961.2 | XP_006253023.1 | 6 | 1106 | |||
XM_006252963.2 | XP_006253025.1 | 3 | 1462 |