Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728731.3 | XP_002728777.1 | 20 | 2108 | ||
XM_008759256.1 | XP_008757478.1 | 20 | 2100 |