Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108488.1 | NP_001101958.1 | 4 | 534 | ||
GenBank mRNA | BC166489.1 | - | 4 | 457 | 7104787 |