Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748924.3 | XP_003748972.1 | 11 | 2142 | ||
XM_006229687.2 | XP_006229749.1 | 10 | 5628 | |||
XM_006229694.2 | XP_006229756.1 | 11 | 2142 | |||
XM_008759651.1 | XP_008757873.1 | 11 | 5768 | |||
XM_008759652.1 | XP_008757874.1 | 11 | 5919 | |||
XM_008759653.1 | XP_008757875.1 | 11 | 8307 | |||
XM_008759656.1 | XP_008757878.1 | 11 | 5768 |