Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748924.3 | XP_003748972.1 | 5 | 924 | ||
XM_006229687.2 | XP_006229749.1 | 4 | 792 | |||
XM_006229694.2 | XP_006229756.1 | 5 | 924 | |||
XM_006229695.2 | XP_006229757.1 | 5 | 925 | |||
XM_008759651.1 | XP_008757873.1 | 5 | 932 | |||
XM_008759652.1 | XP_008757874.1 | 5 | 1083 | |||
XM_008759653.1 | XP_008757875.1 | 5 | 3471 | |||
XM_008759654.1 | XP_008757876.1 | 5 | 932 | |||
XM_008759655.1 | XP_008757877.1 | 5 | 932 | |||
XM_008759656.1 | XP_008757878.1 | 5 | 932 |