Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgapb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271210.1 | NP_001258139.1 | 26 | 3701 | ||
XM_006235415.2 | XP_006235477.1 | 26 | 4110 | |||
XM_006235416.2 | XP_006235478.1 | 25 | 4101 | |||
XM_006235417.2 | XP_006235479.1 | 26 | 4098 | |||
XM_006235418.2 | XP_006235480.1 | 25 | 4089 | |||
XM_006235420.2 | XP_006235482.1 | 24 | 4065 | |||
XM_006235421.2 | XP_006235483.1 | 24 | 4053 | |||
XM_008762357.1 | XP_008760579.1 | 25 | 4125 | |||
XM_008762358.1 | XP_008760580.1 | 24 | 4089 |