Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam76a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108686.1 | NP_001102156.1 | 204 | |||
GenBank mRNA | FQ211538.1 | - | 224 | |||
FQ222884.1 | - | 206 |
Gene Symbol: LOC103690073 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773876.1 | XP_008772098.1 | 285 |