Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191871.1 | NP_001178800.1 | 16, 17 | 3408 | ||
XM_006239232.2 | XP_006239294.1 | 16, 17 | 3443 | |||
XM_006239233.2 | XP_006239295.1 | 16, 17 | 3446 | |||
XM_008764263.1 | XP_008762485.1 | 16, 17 | 3447 | |||
XM_008764264.1 | XP_008762486.1 | 16, 17 | 9005 | |||
XM_008764265.1 | XP_008762487.1 | 17, 18 | 4148 | |||
XM_008764266.1 | XP_008762488.1 | 16, 17 | 8390 | |||
XM_008764267.1 | XP_008762489.1 | 17, 18 | 3537 | |||
XM_008764268.1 | XP_008762490.1 | 15, 16 | 3340 | |||
XM_008764269.1 | XP_008762491.1 | 16, 17 | 3444 |