Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lclat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764536.1 | XP_008762758.1 | 2, 3 | 7764 | ||
XM_008764537.1 | XP_008762759.1 | 2, 3 | 314 | |||
XM_008764538.1 | XP_008762760.1 | 2, 3 | 304 | |||
XM_008764539.1 | XP_008762761.1 | 2, 3 | 7764 |