Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240091.1 | XP_006240153.1 | 28 | 4358 | ||
XM_006240095.1 | XP_006240157.1 | 27 | 4331 | |||
XM_006240096.1 | XP_006240158.1 | 27 | 4328 | |||
XM_008764667.1 | XP_008762889.1 | 28 | 4358 | |||
XM_008764668.1 | XP_008762890.1 | 28 | 4355 |