Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep295 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242549.2 | XP_006242611.2 | 15 | 3459 | ||
XM_008765949.1 | XP_008764171.1 | 15 | 3459 | |||
XM_008765950.1 | XP_008764172.1 | 15 | 3459 | |||
XM_008765951.1 | XP_008764173.1 | 15 | 3459 | |||
XM_008765952.1 | XP_008764174.1 | 15 | 3459 | |||
XM_008765953.1 | XP_008764175.1 | 15 | 3432 | |||
XM_008765954.1 | XP_008764176.1 | 15 | 3460 | |||
XM_008765955.1 | XP_008764177.1 | 15 | 3459 | |||
XM_008765956.1 | XP_008764178.1 | 15 | 3459 | |||
XM_008765957.1 | XP_008764179.1 | 15 | 3459 | |||
GenBank mRNA | EF460314.1 | - | 11 | 2753 |