Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam63b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766411.1 | XP_008764633.1 | 5, 6 | 1442 | ||
XM_343420.8 | XP_343421.3 | 5, 6 | 1434 |